missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NGF, Recombinant Protein
CHO cells Recombinant Protein
70.25 € - 308.00 €
Specifications
Accession Number | P01138 |
---|---|
For Use With (Application) | Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot |
Formulation | 50 mM sodium acetate with 0.02% Tween 80, 150 mM NaCl, 5% mannitol, 5% trehalose and no preservative; pH 5.5 |
Gene ID (Entrez) | 4803 |
Name | Human NGF |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17154243
|
Invitrogen™
A42577 |
5 μg |
70.25 €
5µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17164243
|
Invitrogen™
A42578 |
50 μg |
120.00 €
50µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17134243
|
Invitrogen™
A42575 |
5 x 5 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17144243
|
Invitrogen™
A42576 |
5 x 50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Nerve growth factor (NGF) influences the survival and differentiation of a variety of neuronal and nonneuronal cells. The actions of this polypeptide result from binding to specific cell surface receptors which are present as both high and low affinity sites. Although both types of receptor bind NGF only the high affinity form is capable of signal transduction. Cross-linking studies with 125I-NGF have shown that the high affinity receptor of rat PC12 cells forms an Mr 158,000 complex with NGF while the low affinity receptor forms an Mr 100,000 complex. Full length cDNA clones for both the human and rat NGF receptor genes have been produced and sequenced, revealing greater than 90% homology between the two species. NGF exhibits nerve growth stimulating activity and is involved in the regulation of growth and the differentiation of sympathetic and certain sensory neurons. Further, NGF stimulates division and differentiation of sympathetic and embryonic sensory neurons. Mutations in the NGF gene have been associated with hereditary sensory and autonomic neuropathy, type 5 (HSAN5), and dysregulation of NGF gene expression is associated with allergic rhinitis.Specifications
P01138 | |
50 mM sodium acetate with 0.02% Tween 80, 150 mM NaCl, 5% mannitol, 5% trehalose and no preservative; pH 5.5 | |
Human NGF | |
-20°C | |
<1 EU/μg | |
NGF | |
Unconjugated | |
Lyophilized |
Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot | |
4803 | |
CHO cells | |
RUO | |
B nerve growth factor; beta ngf; beta-nerve growth factor; BetaNGF; beta-NGF; bNGF; b-NGF; HSAN5; MGC161426; MGC161428; nerve growth factor; nerve growth factor (beta polypeptide); nerve growth factor, beta; nerve growth factor, beta subunit; NGF; NGF b; NGF β; NGFB; NGFbeta; NGFβ; β nerve growth factor; β NGF; βNGF | |
Human | |
Recombinant | |
≥ 95% |