missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MYOM3 Partial ORF (NP_689585, 854 a.a. - 955 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00127294-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16101737

  • 508.00 € / 25µg
Estimated Shipment: 27-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: GPYFERPLQWKVTEDCQVQLTCKVTNTKKETRFQWFFQRAEMPDGQYDPETGTGLLCIEELSKKDKGIYRAMVSDDRGEDDTILDLTGDALDAIFTELGRIG
Specifications

Specifications

NP_689585
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.96kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GPYFERPLQWKVTEDCQVQLTCKVTNTKKETRFQWFFQRAEMPDGQYDPETGTGLLCIEELSKKDKGIYRAMVSDDRGEDDTILDLTGDALDAIFTELGRIG
RUO
MYOM3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
127294
MYOM3 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ35961/MGC71483/RP11-293P20.1
MYOM3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions