missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MTL5 Partial ORF (NP_004914, 399 a.a. - 508 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_004914 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9633 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16111086
|
Abnova™
H00009633-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16101086
|
Abnova™
H00009633-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Metallothionein proteins are highly conserved low-molecular-weight cysteine-rich proteins that are induced by and bind to heavy metal ions and have no enzymatic activity. They may play a central role in the regulation of cell growth and differentiation and are involved in spermatogenesis. This gene encodes a metallothionein-like protein which has been shown to be expressed differentially in mouse testis and ovary. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: GCKNYEESPERKTLMSMPNYMQTGGLEGSHYLPPTKFSGLPRFSHDRRPSSCISWEVVEATCACLLAQGEEAEKEHCSKCLAEQMILEEFGRCLSQILHTEFKSKGLKMESpecifications
NP_004914 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CXCDC2/MTLT/TESMIN | |
MTL5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9633 | |
MTL5 (Human) Recombinant Protein (Q01) | |
GCKNYEESPERKTLMSMPNYMQTGGLEGSHYLPPTKFSGLPRFSHDRRPSSCISWEVVEATCACLLAQGEEAEKEHCSKCLAEQMILEEFGRCLSQILHTEFKSKGLKME | |
RUO | |
MTL5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |