missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MRPL50 Full-length ORF (NP_061924.1, 1 a.a. - 158 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00 € - 508.00 €

Specifications

Accession Number NP_061924.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 54534
Molecular Weight (g/mol) 44.7kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16180623
View Documents Promotion Details
Abnova™
H00054534-P01.25UG
25 ug
508.00 €
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 10-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16170623
View Documents Promotion Details
Abnova™
H00054534-P01.10UG
10 ug
335.00 €
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 10-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a putative 39S subunit protein and belongs to the L47P ribosomal protein family. Pseudogenes corresponding to this gene are found on chromosomes 2p, 2q, 5p, and 10q. [provided by RefSeq]

Sequence: MAARSVSGITRRVFMWTVSGTPCREFWSRFRKEKEPVVVETVEEKKEPILVCPPLRSRAYTPPEDLQSRLESYVKEVFGSSLPSNWQDISLEDSRLKFNLLAHLADDLGHVVPNSRLHQMCRVRDVLDFYNVPIQDRSKFDELSASNLPPNLKITWSY
Specifications

Specifications

NP_061924.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
44.7kDa
Glutathione Sepharose 4 Fast Flow
MAARSVSGITRRVFMWTVSGTPCREFWSRFRKEKEPVVVETVEEKKEPILVCPPLRSRAYTPPEDLQSRLESYVKEVFGSSLPSNWQDISLEDSRLKFNLLAHLADDLGHVVPNSRLHQMCRVRDVLDFYNVPIQDRSKFDELSASNLPPNLKITWSY
RUO
MRPL50
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
54534
MRPL50 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ20493/FLJ21990/MRP-L50
MRPL50
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Product Certifications