missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MRPL40 Partial ORF (NP_003767.2, 1 a.a. - 51 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00064976-Q01.10ug

Weitere Details : Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16139776

  • 302.00 € / 10 Mikrogramm


Verfügbar ab
22-09-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Sequence: MTASVLRSISLALRPTSGLLGTWQTQLRETHQRASLLSFWELIPMRSEPLR
Spezifikation

Spezifikation

NP_003767.2
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.35kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MTASVLRSISLALRPTSGLLGTWQTQLRETHQRASLLSFWELIPMRSEPLR
RUO
MRPL40
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
64976
MRPL40 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ41774|MGC9400|MRP-L22|NLVCF|URIM
MRPL40
Yes
Videos
Dokumentation