missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MOGAT2 Partial ORF (NP_079374, 44 a.a. - 106 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_079374 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 80168 |
Molecular Weight (g/mol) | 32.67kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16190187
|
Abnova™
H00080168-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16180187
|
Abnova™
H00080168-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Dietary fat absorption from the small intestine is facilitated by acyl-CoA:monoacylglycerol transferase (MOGAT; EC 2.3.1.22) and acyl-CoA:diacylglycerol acyltransferase (DGAT; see MIM 604900) activities. MOGAT catalyzes the joining of monoacylglycerol and fatty acyl-CoAs to form diacylglycerol (Yen and Farese, 2003 [PubMed 12621063]).[supplied by OMIM]
Sequence: TRFWLLTVLYAAWWYLDRDKPRQGGRHIQAIRCWTIWKYMKDYFPISLVKTAELDPSRNYIAGSpecifications
NP_079374 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.67kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DGAT2L5/FLJ22644/MGAT2/MGC119183/MGC119184/MGC119185 | |
MOGAT2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
80168 | |
MOGAT2 (Human) Recombinant Protein (Q01) | |
TRFWLLTVLYAAWWYLDRDKPRQGGRHIQAIRCWTIWKYMKDYFPISLVKTAELDPSRNYIAG | |
RUO | |
MOGAT2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |