Learn More
Abnova™ Human MOCS2 Full-length ORF (NP_789776.1, 1 a.a. - 88 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004338-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Eukaryotic molybdoenzymes use a unique molybdenum cofactor (MoCo) consisting of a pterin, termed molybdopterin, and the catalytically active metal molybdenum. MoCo is synthesized from precursor Z by the heterodimeric enzyme molybdopterin synthase. The large and small subunits of molybdopterin synthase are both encoded from this gene by overlapping open reading frames. The proteins were initially thought to be encoded from a bicistronic transcript. They are now thought to be encoded from monocistronic transcripts. Alternatively spliced transcripts have been found for this locus that encode the large and small subunits. [provided by RefSeq]
Sequence: MVPLCQVEVLYFAKSAEITGVRSETISVPQEIKALQLWKEIETRHPGLADVRNQIIFAVRQEYVELGDQLLVLQPGDEIAVIPPISGGSpecifications
NP_789776.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MCBPE/MOCO1/MOCS2A/MOCS2B/MPTS | |
MOCS2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
4338 | |
MOCS2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVPLCQVEVLYFAKSAEITGVRSETISVPQEIKALQLWKEIETRHPGLADVRNQIIFAVRQEYVELGDQLLVLQPGDEIAVIPPISGG | |
RUO | |
MOCS2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |