Learn More
Abnova™ Human MLCK Partial ORF (NP_872299.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00091807-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Phosphorylation of cardiac myosin heavy chains (see MYH7B, MIM 609928) and light chains (see MYL2, MIM 160781) by a kinase, such as MYLK3, potentiates the force and rate of cross-bridge recruitment in cardiac myocytes (Chan et al., 2008 [PubMed 18202317]).[supplied by OMIM]
Sequence: MDTKLNMLNEKVDQLLHFQEDVTEKLQSMCRDMGHLERGLHRLEASRAPGPGGADGVPHIDTQAGWPEVLELVRAMQQDAAQHGARLEALFRMVAAVDRASpecifications
NP_872299.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDTKLNMLNEKVDQLLHFQEDVTEKLQSMCRDMGHLERGLHRLEASRAPGPGGADGVPHIDTQAGWPEVLELVRAMQQDAAQHGARLEALFRMVAAVDRA | |
RUO | |
MYLK3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
91807 | |
MLCK (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC126319/MGC126320/MLCK/MLCK2/caMLCK | |
MYLK3 | |
Recombinant | |
wheat germ expression system |