missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MLCK Partial ORF (NP_872299.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00 € - 508.00 €

Specifications

Accession Number NP_872299.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 91807
Molecular Weight (g/mol) 36.74kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16191187
View Documents Promotion Details
Abnova™
H00091807-Q01.25UG
25 ug
508.00 €
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 10-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16181187
View Documents Promotion Details
Abnova™
H00091807-Q01.10UG
10 ug
335.00 €
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 10-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Phosphorylation of cardiac myosin heavy chains (see MYH7B, MIM 609928) and light chains (see MYL2, MIM 160781) by a kinase, such as MYLK3, potentiates the force and rate of cross-bridge recruitment in cardiac myocytes (Chan et al., 2008 [PubMed 18202317]).[supplied by OMIM]

Sequence: MDTKLNMLNEKVDQLLHFQEDVTEKLQSMCRDMGHLERGLHRLEASRAPGPGGADGVPHIDTQAGWPEVLELVRAMQQDAAQHGARLEALFRMVAAVDRA
Specifications

Specifications

NP_872299.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC126319/MGC126320/MLCK/MLCK2/caMLCK
MYLK3
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
91807
MLCK (Human) Recombinant Protein (Q01)
MDTKLNMLNEKVDQLLHFQEDVTEKLQSMCRDMGHLERGLHRLEASRAPGPGGADGVPHIDTQAGWPEVLELVRAMQQDAAQHGARLEALFRMVAAVDRA
RUO
MYLK3
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications