missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MGC34821 Full-length ORF (NP_775857.1, 1 a.a. - 322 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00283238-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16139843

  • 508.00 € / 25µg
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SLC22A24 belongs to a large family of transmembrane proteins that function as uniporters, symporters, and antiporters to transport organic ions across cell membranes (Jacobsson et al., 2007 [PubMed 17714910]).[supplied by OMIM]

Sequence: MGFDVLLDQVGGMGRFQICLIAFFCITNILLFPNIVLENFTAFTPSHRCWVPLLDNDSVSDNDTGTLSKDDLLRISIPLDSNLRPQKCQRFIHPQWQLLHLNGTFPNTNEPDTEPCVDGWVYDRSSFLSTIVTEWDLVCESQSLKSMVQSLFMAGSLLGGLIYGHLSDRVGRKIICKLCFLQLAISNTCAAFAPTFLVYCILRFLAGFSTMTILGNTFILSLEWTLPRSRSMTIMVLLCSYSVGQMLLGGLAFAIQDWHILQLTVSTPIIVLFLSSWYEQSPHSLPVSEAMVDIERKIVTPGICSVSGLVLSHDVHSTYCVT
Specifications

Specifications

NP_775857.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
62.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC34821
SLC22A24
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
283238
MGC34821 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGFDVLLDQVGGMGRFQICLIAFFCITNILLFPNIVLENFTAFTPSHRCWVPLLDNDSVSDNDTGTLSKDDLLRISIPLDSNLRPQKCQRFIHPQWQLLHLNGTFPNTNEPDTEPCVDGWVYDRSSFLSTIVTEWDLVCESQSLKSMVQSLFMAGSLLGGLIYGHLSDRVGRKIICKLCFLQLAISNTCAAFAPTFLVYCILRFLAGFSTMTILGNTFILSLEWTLPRSRSMTIMVLLCSYSVGQMLLGGLAFAIQDWHILQLTVSTPIIVLFLSSWYEQSPHSLPVSEAMVDIERKIVTPGICSVSGLVLSHDVHSTYCVT
RUO
SLC22A24
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions