missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MAGED2 Partial ORF (NP_055414, 16 a.a. - 125 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_055414 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10916 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16193366
|
Abnova™
H00010916-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16183366
|
Abnova™
H00010916-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene is a member of the MAGED gene family. While the MAGEA and MAGEB genes are silent in normal tissues with the exception of testis and placenta, the MAGED genes are expressed ubiquitously. The MAGED genes are clustered on chromosome Xp11. This gene is located in Xp11.2, a hot spot for X-linked mental retardation (XLMR). Multiple alternatively spliced transcript variants have been found for this gene, however, the full length nature of some variants has not been defined. [provided by RefSeq]
Sequence: AEASEKDSSSMMQTLLTVTQNVEVPETPKASKALEVSEDVKVSKASGVSKATEVSKTPEAREAPATQASSTTQLTDTQVLAAENKSLAADTKKQNADPQAVTMPATETKKSpecifications
NP_055414 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
11B6/BCG1/HCA10/JCL-1/MAGE-D2/MAGED/MGC8386 | |
MAGED2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
10916 | |
MAGED2 (Human) Recombinant Protein (Q01) | |
AEASEKDSSSMMQTLLTVTQNVEVPETPKASKALEVSEDVKVSKASGVSKATEVSKTPEAREAPATQASSTTQLTDTQVLAAENKSLAADTKKQNADPQAVTMPATETKK | |
RUO | |
MAGED2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |