missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MAB21L1 Partial ORF (NP_005575, 250 a.a. - 358 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004081-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100855

  • 508.00 € / 25µg
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is similar to the MAB-21 cell fate-determining gene found in C. elegans. It may be involved in eye and cerebellum development, and it has been proposed that expansion of a trinucleotide repeat region in the 5' UTR may play a role in a variety of psychiatric disorders. [provided by RefSeq]

Sequence: CLSILKTLRDRHLELPGQPLNNYHMKTLVSYECEKHPRESDWDESCLGDRLNGILLQLISCLQCRRCPHYFLPNLDLFQGKPHSALENAAKQTWRLAREILTNPKSLEK
Specifications

Specifications

NP_005575
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
CLSILKTLRDRHLELPGQPLNNYHMKTLVSYECEKHPRESDWDESCLGDRLNGILLQLISCLQCRRCPHYFLPNLDLFQGKPHSALENAAKQTWRLAREILTNPKSLEK
RUO
MAB21L1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4081
MAB21L1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAGR1/FLJ10197/Nbla00126
MAB21L1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions