missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KRT6IRS Partial ORF (NP_258259.1, 251 a.a. - 332 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00112802-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16101347

  • 508.00 € / 25µg
Estimated Shipment: 07-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. This gene encodes a protein that is expressed in the inner root sheath of hair follicles. The type II keratins are clustered in a region of chromosome 12q13

Sequence: LQAKVESMDQEIKFFRCLFEAEITQIQSHISDMSVILSMDNNRNLDLDSIIDEVRTQYEEIALKSKAEAEALYQTKFQELQL
Specifications

Specifications

NP_258259.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.76kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LQAKVESMDQEIKFFRCLFEAEITQIQSHISDMSVILSMDNNRNLDLDSIIDEVRTQYEEIALKSKAEAEALYQTKFQELQL
RUO
KRT71
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
112802
KRT6IRS (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
K6IRS1/KRT6IRS/KRT6IRS1/MGC119390/MGC119391
KRT71
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions