missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KHK Partial ORF (NP_000212, 201 a.a. - 297 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003795-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100425

  • 508.00 € / 25µg
Estimated Shipment: 27-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes ketohexokinase that catalyzes conversion of fructose to fructose-1-phosphate. The product of this gene is the first enzyme with a specialized pathway that catabolizes dietary fructose. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: FQSAEEALRGLYGRVRKGAVLVCAWAEEGADALGPDGKLLHSDAFPPPRVVDTLGAGDTFNASVIFSLSQGRSVQEALRFGCQVAGKKCGLQGFDGI
Specifications

Specifications

NP_000212
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.41kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
FQSAEEALRGLYGRVRKGAVLVCAWAEEGADALGPDGKLLHSDAFPPPRVVDTLGAGDTFNASVIFSLSQGRSVQEALRFGCQVAGKKCGLQGFDGI
RUO
KHK
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
3795
KHK (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KHK
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions