missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human IMP3 Full-length ORF (NP_060755.1, 1 a.a. - 184 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00055272-P01.10ug
Weitere Details : Gewicht : 0.00010kg
Beschreibung
Sequence: MVRKLKFHEQKLLKQVDFLNWEVTDHNLHELRVLRRYRLQRREDYTRYNQLSRAVRELARRLRDLPERDQFRVRASAALLDKLYALGLVPTRGSLELCDFVTASSFCRRRLPTVLLKLRMAQHLQAAVAFVEQGHVRVGPDVVTDPAFLVTRSMEDFVTWVDSSKIKRHVLEYNEERDDFDLEASpezifikation
NP_060755.1 | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
48.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BRMS2|C15orf12|DKFZp586L0118|FLJ10968|MRPS4 | |
IMP3 | |
Yes |
wheat germ expression system | |
Liquid | |
55272 | |
IMP3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVRKLKFHEQKLLKQVDFLNWEVTDHNLHELRVLRRYRLQRREDYTRYNQLSRAVRELARRLRDLPERDQFRVRASAALLDKLYALGLVPTRGSLELCDFVTASSFCRRRLPTVLLKLRMAQHLQAAVAFVEQGHVRVGPDVVTDPAFLVTRSMEDFVTWVDSSKIKRHVLEYNEERDDFDLEA | |
RUO | |
IMP3 | |
Wheat Germ (in vitro) | |
GST |