missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human IMP3 Full-length ORF (NP_060755.1, 1 a.a. - 184 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_060755.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 55272 |
Molecular Weight (g/mol) | 48.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16181353
|
Abnova™
H00055272-P01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16191353
|
Abnova™
H00055272-P01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes the human homolog of the yeast Imp3 protein. The protein localizes to the nucleoli and interacts with the U3 snoRNP complex. The protein contains an S4 domain. [provided by RefSeq]
Sequence: MVRKLKFHEQKLLKQVDFLNWEVTDHNLHELRVLRRYRLQRREDYTRYNQLSRAVRELARRLRDLPERDQFRVRASAALLDKLYALGLVPTRGSLELCDFVTASSFCRRRLPTVLLKLRMAQHLQAAVAFVEQGHVRVGPDVVTDPAFLVTRSMEDFVTWVDSSKIKRHVLEYNEERDDFDLEASpecifications
NP_060755.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
48.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MVRKLKFHEQKLLKQVDFLNWEVTDHNLHELRVLRRYRLQRREDYTRYNQLSRAVRELARRLRDLPERDQFRVRASAALLDKLYALGLVPTRGSLELCDFVTASSFCRRRLPTVLLKLRMAQHLQAAVAFVEQGHVRVGPDVVTDPAFLVTRSMEDFVTWVDSSKIKRHVLEYNEERDDFDLEA | |
RUO | |
IMP3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
55272 | |
IMP3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BRMS2/C15orf12/DKFZp586L0118/FLJ10968/MRPS4 | |
IMP3 | |
Recombinant | |
wheat germ expression system |