missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HSD17B6 Partial ORF (NP_003716.2, 180 a.a. - 280 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00008630-Q01.10ug

Weitere Details : Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16138915

  • 302.00 € / 10 Mikrogramm


Verfügbar ab
25-09-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Sequence: KYGVEAFSDILRREIQHFGVKISIVEPGYFRTGMTNMTQSLERMKQSWKEAPKHIKETYGQQYFDALYNIMKEGLLNCSTNLNLVTDCMEHALTSVHPRTR
Spezifikation

Spezifikation

NP_003716.2
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.85kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KYGVEAFSDILRREIQHFGVKISIVEPGYFRTGMTNMTQSLERMKQSWKEAPKHIKETYGQQYFDALYNIMKEGLLNCSTNLNLVTDCMEHALTSVHPRTR
RUO
HSD17B6
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
8630
HSD17B6 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HSE|RODH|SDR9C6
HSD17B6
Yes
Videos
Dokumentation