missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HMGCS2 Partial ORF (NP_005509.1, 424 a.a. - 508 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003158-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16139394

  • 335.00 € / 10µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Mitochondrial 3-hydroxy-3-methylglutaryl CoA synthase (HMGCS2; EC 2.3.3.10) mediates the first reaction of ketogenesis, a metabolic pathway that provides lipid-derived energy for brain, heart, kidney, and other organs during times of carbohydrate deprivation such as fasting (Robinson and Williamson, 1980 [PubMed 6986618]). Also see cytoplasmic HMG-CoA synthase (HMGCS1; MIM 142940), which mediates an early step in cholesterol synthesis.[supplied by OMIM]

Sequence: RVSQDAAPGSPLDKLVSSTSDLPKRLASRKCVSPEEFTEIMNQREQFYHKVNFSPPGDTNSLFPGTWYLERVDEQHRRKYARRPV
Specifications

Specifications

NP_005509.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.09kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RVSQDAAPGSPLDKLVSSTSDLPKRLASRKCVSPEEFTEIMNQREQFYHKVNFSPPGDTNSLFPGTWYLERVDEQHRRKYARRPV
RUO
HMGCS2
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
3158
HMGCS2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HMGCS2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions