missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HMGCS2 Full-length ORF (NP_005509.1, 1 a.a. - 508 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
POA
Specifications
Name | HMGCS2 (Human) Recombinant Protein (P01) |
---|---|
Recombinant | Yes |
Purification Method | Glutathione Sepharose 4 Fast Flow |
Molecular Weight (g/mol) | 83kDa |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Description
Mitochondrial 3-hydroxy-3-methylglutaryl CoA synthase (HMGCS2; EC 2.3.3.10) mediates the first reaction of ketogenesis, a metabolic pathway that provides lipid-derived energy for brain, heart, kidney, and other organs during times of carbohydrate deprivation such as fasting (Robinson and Williamson, 1980 [PubMed 6986618]). Also see cytoplasmic HMG-CoA synthase (HMGCS1; MIM 142940), which mediates an early step in cholesterol synthesis.[supplied by OMIM]
Sequence: MQRLLTPVKRILQLTRAVQETSLTPARLLPVAHQRFSTASAVPLAKTDTWPKDVGILALEVYFPAQYVDQTDLEKYNNVEAGKYTVGLGQTRMGFCSVQEDINSLCLTVVQRLMERIQLPWDSVGRLEVGTETIIDKSKAVKTVLMELFQDSGNTDIEGIDTTNACYGGTASLFNAANWMESSSWDGRYAMVVCGDIAVYPSGNARPTGGAGAVAMLIGPKAPLALERGLRGTHMENVYDFYKPNLASEYPIVDGKLSIQCYLRALDRCYTSYRKKIQNQWKQAGSDRPFTLDDLQYMIFHTPFCKMVQKSLARLMFNDFLSASSDTQTSLYKGLEAFGGLKLEDTYTNKDLDKALLKASQDMFDKKTKASLYLSTHNGNMYTSSLYGCLASLLSHHSAQELAGSRIGAFSYGSGLAASFFSFRVSQDAAPGSPLDKLVSSTSDLPKRLASRKCVSPEEFTEIMNQREQFYHKVNFSPPGDTNSLFPGTWYLERVDEQHRRKYARRPVSpecifications
HMGCS2 (Human) Recombinant Protein (P01) | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Yes | |
83kDa |