missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GRWD1 Full-length ORF (AAH02440.1, 1 a.a. - 446 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00083743-P01.25ug

Weitere Details : Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16185463

  • 467.00 € / 25 Mikrogramm


Verfügbar ab
15-02-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Sequence: MAARKGRRRTCETGEPMEAESGDTSSEGPAQVYLPGRGPPLREGEELVMDEEAYVLYHRAQTGAPCLSFDIVRDHLGDNRTELPLTLYLCAGTQAESAQSNRLMMLRMHNLHGTKPPPSEGSDEEEEEEDEEDEEERKPQLELAMVPHYGGINRVRVSWLGEEPVAGVWSEKGQVEVFALRRLLQVVEEPQALAAFLRDEQAQMKPIFSFAGHMGEGFALDWSPRVTGRLLTGDCQKNIHLWTPTDGGSWHVDQRPFVGHTRSVEDLQWSPTENTVFASCSADASIRIWDIRAAPSKACMLTTATAHDGDVNVISWSRREPFLLSGGDDGALKIWDLRQFKSGSPVATFKQHVAPVTSVEWHPQDSGVFAASGADHQITQWDLAVERDPEAGDVEADPGLADLPQQLLFVHQGETELKELHWHPQCPGLLVSTALSGFTIFRTISV
Spezifikation

Spezifikation

AAH02440.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
75.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GRWD|KIAA1942|RRB1|WDR28
GRWD1
Yes
wheat germ expression system
Liquid
83743
GRWD1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAARKGRRRTCETGEPMEAESGDTSSEGPAQVYLPGRGPPLREGEELVMDEEAYVLYHRAQTGAPCLSFDIVRDHLGDNRTELPLTLYLCAGTQAESAQSNRLMMLRMHNLHGTKPPPSEGSDEEEEEEDEEDEEERKPQLELAMVPHYGGINRVRVSWLGEEPVAGVWSEKGQVEVFALRRLLQVVEEPQALAAFLRDEQAQMKPIFSFAGHMGEGFALDWSPRVTGRLLTGDCQKNIHLWTPTDGGSWHVDQRPFVGHTRSVEDLQWSPTENTVFASCSADASIRIWDIRAAPSKACMLTTATAHDGDVNVISWSRREPFLLSGGDDGALKIWDLRQFKSGSPVATFKQHVAPVTSVEWHPQDSGVFAASGADHQITQWDLAVERDPEAGDVEADPGLADLPQQLLFVHQGETELKELHWHPQCPGLLVSTALSGFTIFRTISV
RUO
GRWD1
Wheat Germ (in vitro)
GST
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation