Learn More
Abnova™ Human GRM7 Partial ORF (NP_000835, 431 a.a. - 520 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_000835 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 2917 |
Molecular Weight (g/mol) | 35.64kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16185781
|
Abnova™
H00002917-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16175781
|
Abnova™
H00002917-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
L-glutamate is the major excitatory neurotransmitter in the central nervous system, and it activates both ionotropic and metabotropic glutamate receptors. Glutamatergic neurotransmission is involved in most aspects of normal brain function and can be perturbed in many neuropathologic conditions. The metabotropic glutamate receptors are a family of G protein-coupled receptors that have been divided into three groups on the basis of sequence homology, putative signal transduction mechanisms, and pharmacologic properties. Group I includes GRM1 and GRM5, and these receptors have been shown to activate phospholipase C. Group II includes GRM2 and GRM3, while Group III includes GRM4, GRM6, GRM7 and GRM8. Group II and III receptors are linked to the inhibition of the cyclic AMP cascade but differ in their agonist selectivities. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: ADYRGVCPEMEQAGGKKLLKYIRNVNFNGSAGTPVMFNKNGDAPGRYDIFQYQTTNTSNPGYRLIGQWTDELQLNIEDMQWGKGVREIPASpecifications
NP_000835 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ40498/GLUR7/GPRC1G/MGLUR7/mGlu7 | |
GRM7 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
2917 | |
GRM7 (Human) Recombinant Protein (Q01) | |
ADYRGVCPEMEQAGGKKLLKYIRNVNFNGSAGTPVMFNKNGDAPGRYDIFQYQTTNTSNPGYRLIGQWTDELQLNIEDMQWGKGVREIPA | |
RUO | |
GRM7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |