missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GRCC9 Partial ORF (AAH02983, 1 a.a. - 90 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00 € - 508.00 €

Specifications

Accession Number AAH02983
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 84727
Molecular Weight (g/mol) 35.64kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16110877
View Documents Promotion Details
Abnova™
H00084727-Q01.25UG
25 ug
508.00 €
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 10-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16100877
View Documents Promotion Details
Abnova™
H00084727-Q01.10UG
10 ug
335.00 €
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 10-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene encodes encodes a suppressor of cytokine signaling (SOCS) family member, and it belongs to the subfamily of proteins containing a central SPRY (repeats in splA and RyR) domain and a C-terminal SOCS box. This gene is present in a gene-rich cluster on chromosome 12p13 in the vicinity of the CD4 antigen and triosephosphate isomerase genes. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: MGQTALAGGSSSTPTPQALYPDLSCPEGLEELLSAPPPDLGAQRRHGWNPKDCSENIEVKEGGLYFERRPVAQSTDGARGKRGYSRGLHA
Specifications

Specifications

AAH02983
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.64kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GRCC9/MGC2519/SSB-2/SSB2
SPSB2
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
84727
GRCC9 (Human) Recombinant Protein (Q01)
MGQTALAGGSSSTPTPQALYPDLSCPEGLEELLSAPPPDLGAQRRHGWNPKDCSENIEVKEGGLYFERRPVAQSTDGARGKRGYSRGLHA
RUO
SPSB2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications