missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GPR82 Partial ORF (NP_543007.1, 237 a.a. - 336 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00027197-Q01.10ug
Weitere Details : Gewicht : 0.00010kg
Beschreibung
Sequence: IMEKDLTYSSVKRHLLVIQILLIVCFLPYSIFKPIFYVLHQRDNCQQLNYLIETKNILTCLASARSSTDPIIFLLLDKTFKKTLYNLFTKSNSAHMQSYGSpezifikation
NP_543007.1 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
IMEKDLTYSSVKRHLLVIQILLIVCFLPYSIFKPIFYVLHQRDNCQQLNYLIETKNILTCLASARSSTDPIIFLLLDKTFKKTLYNLFTKSNSAHMQSYG | |
RUO | |
GPR82 | |
Yes |
wheat germ expression system | |
Liquid | |
27197 | |
GPR82 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GPR82 | |
Wheat Germ (in vitro) | |
GST |