missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GNRH1 Partial ORF (NP_000816, 24 a.a. - 92 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00002796-Q01.25ug
Weitere Details : Gewicht : 0.00010kg
Beschreibung
Sequence: QHWSYGLRPGGKRDAENLIDSFQEIVKEVGQLAETQRFECTTHQPRSPLRDLKGALESLIEEETGQKKISpezifikation
NP_000816 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.33kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QHWSYGLRPGGKRDAENLIDSFQEIVKEVGQLAETQRFECTTHQPRSPLRDLKGALESLIEEETGQKKI | |
RUO | |
GNRH1 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
2796 | |
GNRH1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GNRH|GRH|LHRH|LNRH | |
GNRH1 | |
Yes |