Learn More
Abnova™ Human GNA14 Partial ORF (AAH27886.1, 150 a.a. - 250 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009630-Q01.25ug
Additional Details : Weight : 0.02000kg
Description
This gene encodes a member of the guanine nucleotide-binding, or G protein family. G proteins are heterotrimers consisting of alpha, beta and gamma subunits. The encoded protein is a member of the alpha family of G proteins, more specifically the alpha q subfamily of G proteins. The encoded protein may play a role in pertussis-toxin resistant activation of phospholipase C-beta and its downstream effectors
Sequence: SDSAKYYLTDIDRIATPSFVPTQQDVLRVRVPTTGIIEYPFDLENIIFRMVDVGGQRSERRKWIHCFESVTSIIFLVALSEYDQVLAECDNENRMEESKALSpecifications
AAH27886.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SDSAKYYLTDIDRIATPSFVPTQQDVLRVRVPTTGIIEYPFDLENIIFRMVDVGGQRSERRKWIHCFESVTSIIFLVALSEYDQVLAECDNENRMEESKAL | |
RUO | |
GNA14 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9630 | |
GNA14 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GNA14 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |