Learn More
Abnova™ Human GIMAP7 Full-length ORF (NP_694968.1, 1 a.a. - 300 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00168537-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, the IAN subfamily genes are located in a cluster at 7q36.1. [provided by RefSeq]
Sequence: MAESEDRSLRIVLVGKTGSGKSATANTILGEEIFDSRIAAQAVTKNCQKASREWQGRDLLVVDTPGLFDTKESLDTTCKEISRCIISSCPGPHAIVLVLLLGRYTEEEQKTVALIKAVFGKSAMKHMVILFTRKEELEGQSFHDFIADADVGLKSIVKECGNRCCAFSNSKKTSKAEKESQVQELVELIEKMVQCNEGAYFSDDIYKDTEERLKQREEVLRKIYTDQLNEEIKLVEEDKHKSEEEKEKEIKLLKLKYDEKIKNIREEAERNIFKDVFNRIWKMLSEIWHRFLSKCKFYSSSpecifications
NP_694968.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
60.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IAN7/MGC27027/hIAN7 | |
GIMAP7 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
168537 | |
GIMAP7 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAESEDRSLRIVLVGKTGSGKSATANTILGEEIFDSRIAAQAVTKNCQKASREWQGRDLLVVDTPGLFDTKESLDTTCKEISRCIISSCPGPHAIVLVLLLGRYTEEEQKTVALIKAVFGKSAMKHMVILFTRKEELEGQSFHDFIADADVGLKSIVKECGNRCCAFSNSKKTSKAEKESQVQELVELIEKMVQCNEGAYFSDDIYKDTEERLKQREEVLRKIYTDQLNEEIKLVEEDKHKSEEEKEKEIKLLKLKYDEKIKNIREEAERNIFKDVFNRIWKMLSEIWHRFLSKCKFYSS | |
RUO | |
GIMAP7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |