missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FUNDC2 Partial ORF (NP_076423.2, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00065991-Q04.10ug
Additional Details : Weight : 0.02000kg
Description
Sequence: METSAPRAGSQVVATTARHSAAYRADPLRVSSRDKLTEMAASSQGNFEGNFESLDLAEFAKKQPWWRKLFGQESGPSAEKYSVATQLFIGGVTGWCTGFISpecifications
NP_076423.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
METSAPRAGSQVVATTARHSAAYRADPLRVSSRDKLTEMAASSQGNFEGNFESLDLAEFAKKQPWWRKLFGQESGPSAEKYSVATQLFIGGVTGWCTGFI | |
RUO | |
FUNDC2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
65991 | |
FUNDC2 (Human) Recombinant Protein (Q04) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DC44/FLJ33773/HCBP6/HCC3/MGC131676/MGC2495/PD03104 | |
FUNDC2 | |
Recombinant | |
wheat germ expression system |