missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FRS2 Partial ORF (AAH21562.1, 1 a.a. - 341 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010818-Q01.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16075275

  • 518.16 € / 25µg

Estimated Shipment: 14-01-2022
Add to basket

Description

Description

Sequence: MGSCCSCPDKDTVPDNHRNKFKVINVDDDGNELGSGIMELTDTELILYTRKRDSVKWHYLCLRRYGYDSNLFSFESGRRCQTGQGIFAFKCARAEELFNMSQEIMQNNSINVVEEPVVERNNHQTELEVPRTPRTPTTPGFAAQNLPNGYPRYPSFGDASSHPSSRHPSVGSARLPSVGEESTHPLLVAEEQVHTYVNTTGVQEERKNRTSVHVPLEARVSNAESSTPKEEPSSIEDRDPQILLEPEGVKFVLGPTPVQKQLMEKEKLEQLGRDQVSGSGANNTEWDTGYDSDERRDAPSVNKLVYENINGLSIPSASGVRRGRLTSTSTSDTQNINNSAQ
Specifications

Specifications

AAH21562.1
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
63.14kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MGSCCSCPDKDTVPDNHRNKFKVINVDDDGNELGSGIMELTDTELILYTRKRDSVKWHYLCLRRYGYDSNLFSFESGRRCQTGQGIFAFKCARAEELFNMSQEIMQNNSINVVEEPVVERNNHQTELEVPRTPRTPTTPGFAAQNLPNGYPRYPSFGDASSHPSSRHPSVGSARLPSVGEESTHPLLVAEEQVHTYVNTTGVQEERKNRTSVHVPLEARVSNAESSTPKEEPSSIEDRDPQILLEPEGVKFVLGPTPVQKQLMEKEKLEQLGRDQVSGSGANNTEWDTGYDSDERRDAPSVNKLVYENINGLSIPSASGVRRGRLTSTSTSDTQNINNSAQ
RUO
FRS2
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
10818
FRS2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FRS2A/FRS2alpha/SNT/SNT-1/SNT1
FRS2
Yes
Videos
SDS
Documents
Certificates
Promotions

Promotions