Learn More
Abnova™ Human FBXO11 Partial ORF (NP_079409, 744 a.a. - 843 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00080204-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: KAVSRGQCLYKISSYTSYPMHDFYRCHTCNTTDRNAICVNCIKKCHQGHDVEFIRHDRFFCDCGAGTLSNPCTLAGEPTHDTDTLYDSAPPIESNTLQHNSpecifications
NP_079409 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KAVSRGQCLYKISSYTSYPMHDFYRCHTCNTTDRNAICVNCIKKCHQGHDVEFIRHDRFFCDCGAGTLSNPCTLAGEPTHDTDTLYDSAPPIESNTLQHN | |
RUO | |
FBXO11 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
80204 | |
FBXO11 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FBX11/FLJ12673/MGC44383/PRMT9/UBR6/UG063H01/VIT1 | |
FBXO11 | |
Recombinant | |
wheat germ expression system |