Learn More
Abnova™ Human FAM12A Full-length ORF (NP_006674.2, 1 a.a. - 147 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_006674.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10876 |
Molecular Weight (g/mol) | 44kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16175782
|
Abnova™
H00010876-P01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16185782
|
Abnova™
H00010876-P01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Testicular sperm are morphologically differentiated but are not progressively motile nor able to fertilize an egg. Post-testicular maturation requires exposure of spermatozoa to the microenvironment of the epididymal lumen. Spermatozoa undergo extensive changes in the epididymis, including enzymatic modifications, loss of pre-existing components and addition of new glycoproteins from epididymal secretions. These modifying proteins and enzymes are synthesized by epithelial cells lining the epididymal duct and secreted apically into the lumen, where they come into contact with, and may be absorbed onto, the sperm membranes. The proteins encoded by the genes in this cluster are synthesized and secreted by epididymal epithelial cells. [provided by RefSeq]
Sequence: MTSSLKIWGILLALLCILCRLCVYSNNIYWREFIKLHYLSPSREFKEYKCDVLMREKEALKGKSFHMFIYSLWFKIQRACINEKGSDRYRNAYVWAPGALKVLECHWEKYNNRYTESRSFSYIEFHCGVDGYVDNIEDLRIIEPISNSpecifications
NP_006674.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
44kDa | |
Glutathione Sepharose 4 Fast Flow | |
MTSSLKIWGILLALLCILCRLCVYSNNIYWREFIKLHYLSPSREFKEYKCDVLMREKEALKGKSFHMFIYSLWFKIQRACINEKGSDRYRNAYVWAPGALKVLECHWEKYNNRYTESRSFSYIEFHCGVDGYVDNIEDLRIIEPISN | |
RUO | |
FAM12A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10876 | |
FAM12A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EP3A/HE3-ALPHA/HE3A/HE3ALPHA/MGC119614/MGC119615 | |
FAM12A | |
Recombinant | |
wheat germ expression system |