missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ETS2 Partial ORF (AAH42954, 178 a.a. - 277 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00002114-Q01.10ug

Weitere Details : Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16184691

  • 310.00 € / 10 Mikrogramm


Verfügbar ab
17-02-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Sequence: YEENSHLTSVPHWINSNTLGFGTEQAPYGMQTQNYPKGGLLDSMCPASTPSVLSSEQEFQMFPKSRLSSVSVTYCSVSQDFPGSNLNLLTNNSGTPKDHD
Spezifikation

Spezifikation

AAH42954
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
YEENSHLTSVPHWINSNTLGFGTEQAPYGMQTQNYPKGGLLDSMCPASTPSVLSSEQEFQMFPKSRLSSVSVTYCSVSQDFPGSNLNLLTNNSGTPKDHD
RUO
ETS2
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
2114
ETS2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ETS2IT1
ETS2
Yes
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation