missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human EIF3S5 Partial ORF (NP_003745, 258 a.a. - 357 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00008665-Q01.25ug

Weitere Details : Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16138995

  • 458.00 € / 25 Mikrogramm


Verfügbar ab
30-09-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Sequence: SPNRVIGLSSDLQQVGGASARIQDALSTVLQYAEDVLSGKVSADNTVGRFLMSLVNQVPKIVPDDFETMLNSNINDLLMVTYLANLTQSQIALNEKLVNL
Spezifikation

Spezifikation

NP_003745
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SPNRVIGLSSDLQQVGGASARIQDALSTVLQYAEDVLSGKVSADNTVGRFLMSLVNQVPKIVPDDFETMLNSNINDLLMVTYLANLTQSQIALNEKLVNL
RUO
EIF3F
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
8665
EIF3S5 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EIF3S5|eIF3-p47
EIF3F
Yes
Videos
Dokumentation