missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EFS Full-length ORF (NP_115835.1, 1 a.a. - 468 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_115835.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10278 |
Molecular Weight (g/mol) | 75.4kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16184802
|
Abnova™
H00010278-P01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16174802
|
Abnova™
H00010278-P01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene contains a SH3 domain, which is known to be important in intracellular signal transduction. The protein encoded by a similiar gene in mice was shown to be able to bind to SH3 domain of protein-tyrosine kinases. The function of this gene is unknown. Two alternatively spliced variants have been described. [provided by RefSeq]
Sequence: MAIATSVYVVPPPARPCPTSGPPAGPCPPSPDLIYKIPRASGTQLAAPRDALEVYDVPPTALRVPSSGPYDCPASFSHPLTRVAPQPPGEDDAPYDVPLTPKPPAELEPDLEWEGGREPGPPIYAAPSNLKRASALLNLYEAPEELLADGEGGGTDEGIYDVPLLGPEAPPSPEPPGALASHDQDTLAQLLARSPPPPHRPRLPSAESLSRRPLPALPVPEAPSPSPVPSPAPGRKGSIQDRPLPPPPPRLPGYGGPKVEGDPEGREMEDDPAGHHNEYEGIPMAEEYDYVHLKGMDKAQGSRPPDQACTGDPELPERGMPAPQEALSPGEPLVVSTGDLQLLYFYAGQCQSHYSALQAAVAALMSSTQANQPPRLFVPHSKRVVVAAHRLVFVGDTLGRLAASAPLRAQVRAAGTALGQALRATVLAVKGAALGYPSSPAIQEMVQCVTELAGQALQFTTLLTSLAPSpecifications
NP_115835.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
75.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAIATSVYVVPPPARPCPTSGPPAGPCPPSPDLIYKIPRASGTQLAAPRDALEVYDVPPTALRVPSSGPYDCPASFSHPLTRVAPQPPGEDDAPYDVPLTPKPPAELEPDLEWEGGREPGPPIYAAPSNLKRASALLNLYEAPEELLADGEGGGTDEGIYDVPLLGPEAPPSPEPPGALASHDQDTLAQLLARSPPPPHRPRLPSAESLSRRPLPALPVPEAPSPSPVPSPAPGRKGSIQDRPLPPPPPRLPGYGGPKVEGDPEGREMEDDPAGHHNEYEGIPMAEEYDYVHLKGMDKAQGSRPPDQACTGDPELPERGMPAPQEALSPGEPLVVSTGDLQLLYFYAGQCQSHYSALQAAVAALMSSTQANQPPRLFVPHSKRVVVAAHRLVFVGDTLGRLAASAPLRAQVRAAGTALGQALRATVLAVKGAALGYPSSPAIQEMVQCVTELAGQALQFTTLLTSLAP | |
RUO | |
EFS | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10278 | |
EFS (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAS3/CASS3/EFS1/EFS2/HEFS/SIN | |
EFS | |
Recombinant | |
wheat germ expression system |