missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DTYMK Partial ORF (NP_036277, 103 a.a. - 212 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00001841-Q01.25ug
Weitere Details : Gewicht : 0.00010kg
Beschreibung
Sequence: VAFTGAKENFSLDWCKQPDVGLPKPDLVLFLQLQLADAAKRGAFGHERYENGAFQERALRCFHQLMKDTTLNWKMVDASKSIEAVHEDIRVLSEDAIRTATEKPLGELWKSpezifikation
NP_036277 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VAFTGAKENFSLDWCKQPDVGLPKPDLVLFLQLQLADAAKRGAFGHERYENGAFQERALRCFHQLMKDTTLNWKMVDASKSIEAVHEDIRVLSEDAIRTATEKPLGELWK | |
RUO | |
DTYMK | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
1841 | |
DTYMK (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDC8|FLJ44192|TMPK|TYMK | |
DTYMK | |
Yes |