missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DOK6 Full-length ORF (NP_689934.2, 1 a.a. - 331 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00220164-P01.25ug

Weitere Details : Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16139423

  • 458.00 € / 25 Mikrogramm


Verfügbar ab
22-09-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Sequence: MASNFNDIVKQGYVKIRSRKLGIFRRCWLVFKKASSKGPRRLEKFPDEKAAYFRNFHKVTELHNIKNITRLPRETKKHAVAIIFHDETSKTFACESELEAEEWCKHLCMECLGTRLNDISLGEPDLLAAGVQREQNERFNVYLMPTPNLDIYGECTMQITHENIYLWDIHNAKVKLVMWPLSSLRRYGRDSTWFTFESGRMCDTGEGLFTFQTREGEMIYQKVHSATLAIAEQHERLMLEMEQKARLQTSLTEPMTLSKSISLPRSAYWHHITRQNSVGEIYSLQGHGFGSSKMSRAQTFPSYAPEQSEEAQQPLSRSSSYGFSYSSSLIQ
Spezifikation

Spezifikation

NP_689934.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
64.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DOK5L|HsT3226|MGC20785
DOK6
Yes
wheat germ expression system
Liquid
220164
DOK6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASNFNDIVKQGYVKIRSRKLGIFRRCWLVFKKASSKGPRRLEKFPDEKAAYFRNFHKVTELHNIKNITRLPRETKKHAVAIIFHDETSKTFACESELEAEEWCKHLCMECLGTRLNDISLGEPDLLAAGVQREQNERFNVYLMPTPNLDIYGECTMQITHENIYLWDIHNAKVKLVMWPLSSLRRYGRDSTWFTFESGRMCDTGEGLFTFQTREGEMIYQKVHSATLAIAEQHERLMLEMEQKARLQTSLTEPMTLSKSISLPRSAYWHHITRQNSVGEIYSLQGHGFGSSKMSRAQTFPSYAPEQSEEAQQPLSRSSSYGFSYSSSLIQ
RUO
DOK6
Wheat Germ (in vitro)
GST
Videos
Dokumentation