missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DNAJB8 Full-length ORF (NP_699161.1, 1 a.a. - 232 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_699161.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 165721 |
Molecular Weight (g/mol) | 52.1kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16139023
|
Abnova™
H00165721-P01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16129023
|
Abnova™
H00165721-P01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
DNAJB8 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain (Ohtsuka and Hata, 2000 [PubMed 11147971]).[supplied by OMIM]
Sequence: MANYYEVLGVQASASPEDIKKAYRKLALRWHPDKNPDNKEEAEKKFKLVSEAYEVLSDSKKRSLYDRAGCDSWRAGGGASTPYHSPFDTGYTFRNPEDIFREFFGGLDPFSFEFWDSPFNSDRGGRGHGLRGAFSAGFGEFPAFMEAFSSFNMLGCSGGSHTTFSSTSFGGSSSGSSGFKSVMSSTEMINGHKVTTKRIVENGQERVEVEEDGQLKSVTVNGKEQLKWMDSKSpecifications
NP_699161.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
52.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
MANYYEVLGVQASASPEDIKKAYRKLALRWHPDKNPDNKEEAEKKFKLVSEAYEVLSDSKKRSLYDRAGCDSWRAGGGASTPYHSPFDTGYTFRNPEDIFREFFGGLDPFSFEFWDSPFNSDRGGRGHGLRGAFSAGFGEFPAFMEAFSSFNMLGCSGGSHTTFSSTSFGGSSSGSSGFKSVMSSTEMINGHKVTTKRIVENGQERVEVEEDGQLKSVTVNGKEQLKWMDSK | |
RUO | |
DNAJB8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
165721 | |
DNAJB8 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DJ6/MGC33884 | |
DNAJB8 | |
Recombinant | |
wheat germ expression system |