missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DENND1B Full-length ORF (NP_659414.1, 1 a.a. - 396 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00163486-P01.10ug

Weitere Details : Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16138973

  • 302.00 € / 10 Mikrogramm


Verfügbar ab
23-09-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Sequence: MAAAPREEKRWPQPVFSNPVVLWKFPEDFGDQEILQSVPKFCFPFDVERVSQNQVGQHFTFVLTDIESKQRFGFCRLTSGGTICLCILSYLPWFEVYYKLLNTLADYLAKELENDLNETLRSLYNHPVPKANTPVNLSVHSYFIAPDVTGLPTIPESRNLTEYFVAVDVNNMLQLYASMLHERRIVIISSKLSTLTACIHGSAALLYPMYWQHIYIPVLPPHLLDYCCAPMPYLIGIHSSLIERVKNKSLEDVVMLNVDTNTLESPFSDLNNLPSDVVSALKNKLKKQSTATGDGVARAFLRAQAALFGYYRDALRYKPGEPITFCEESFVKHRSSVMKQFLETAINLQLFKQFIDGRLAKLNAGRGFSDVFEEEITSGGFCGGKDKLQYDYPFSQ
Spezifikation

Spezifikation

NP_659414.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
71.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C1orf218|DKFZp547O0715|FAM31B|FLJ20054|FLJ21179|FLJ33888|MGC103810|MGC27044
DENND1B
Yes
wheat germ expression system
Liquid
163486
DENND1B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAAPREEKRWPQPVFSNPVVLWKFPEDFGDQEILQSVPKFCFPFDVERVSQNQVGQHFTFVLTDIESKQRFGFCRLTSGGTICLCILSYLPWFEVYYKLLNTLADYLAKELENDLNETLRSLYNHPVPKANTPVNLSVHSYFIAPDVTGLPTIPESRNLTEYFVAVDVNNMLQLYASMLHERRIVIISSKLSTLTACIHGSAALLYPMYWQHIYIPVLPPHLLDYCCAPMPYLIGIHSSLIERVKNKSLEDVVMLNVDTNTLESPFSDLNNLPSDVVSALKNKLKKQSTATGDGVARAFLRAQAALFGYYRDALRYKPGEPITFCEESFVKHRSSVMKQFLETAINLQLFKQFIDGRLAKLNAGRGFSDVFEEEITSGGFCGGKDKLQYDYPFSQ
RUO
DENND1B
Wheat Germ (in vitro)
GST
Videos
Dokumentation