Learn More
Abnova™ Human DAZ4 Full-length ORF (NP_065153.1, 1 a.a. - 390 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00057135-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the DAZ gene family and is a candidate for the human Y-chromosomal azoospermia factor (AZF). Its expression is restricted to premeiotic germ cells, particularly in spermatogonia. It encodes an RNA-binding protein that is important for spermatogenesis. Four copies of this gene are found on chromosome Y within palindromic duplications; one pair of genes is part of the P2 palindrome and the second pair is part of the P1 palindrome. Each gene contains a 2.4 kb repeat including a 72-bp exon, called the DAZ repeat; the number of DAZ repeats is variable and there are several variations in the sequence of the DAZ repeat. Each copy of the gene also contains a 10.8 kb region that may be amplified; this region includes five exons that encode an RNA recognition motif (RRM) domain. This gene contains two copies of the 10.8 kb repeat. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: MSAANPETPNSTISREASTQSSSAAASQGWVLPEGKIVPNTVFVGGIDARMDETEIGSCFGRYGSVKEVKIITNRTGVSKGYGFVSFVNDVDVQKIVGSQIHFHGKKLKLGPAIRKQKLCARHVQPRPLVVNPPPPPQFQNVWRNPNTETYLQPQITPNPVTQHVQAYSAYPHSPGQVITGCQLLVYNYQEYPTYPDSAFQVTTGYQLPVYNYQPFPAYPRSPFQVTAGYQLPVYNYQAFPAYPNSPFQVATGYQFPVYNYQPFPAYPSSPFQVTAGYQLPVYNYQAFPAYPNSPFQVATGYQFPVYNYQAFPAYPNSPVQVTTGYQLPVYNYQAFPAYPNSAVQVTTGYQFHVYNYQMPPQCPVGEQRRNLWTEAYKWWYLVCLIQRRDSpecifications
NP_065153.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
70.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DAZ/DAZ1/pDP1680/pDP1681 | |
DAZ4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
57135 | |
DAZ4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSAANPETPNSTISREASTQSSSAAASQGWVLPEGKIVPNTVFVGGIDARMDETEIGSCFGRYGSVKEVKIITNRTGVSKGYGFVSFVNDVDVQKIVGSQIHFHGKKLKLGPAIRKQKLCARHVQPRPLVVNPPPPPQFQNVWRNPNTETYLQPQITPNPVTQHVQAYSAYPHSPGQVITGCQLLVYNYQEYPTYPDSAFQVTTGYQLPVYNYQPFPAYPRSPFQVTAGYQLPVYNYQAFPAYPNSPFQVATGYQFPVYNYQPFPAYPSSPFQVTAGYQLPVYNYQAFPAYPNSPFQVATGYQFPVYNYQAFPAYPNSPVQVTTGYQLPVYNYQAFPAYPNSAVQVTTGYQFHVYNYQMPPQCPVGEQRRNLWTEAYKWWYLVCLIQRRD | |
RUO | |
DAZ4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |