missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CYP11A1 Partial ORF (AAH32329, 412 a.a. - 521 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00001583-Q01.25ug
Weitere Details : Gewicht : 0.00010kg
Beschreibung
Sequence: KTLVQVAIYALGREPTFFFDPENFDPTRWLSKDKNITYFRNLGFGWGVRQCLGRRIAELEMTIFLINMLENFRVEIQHLSDVGTTFNLILMPEKPISFTFWPFNQEATQQSpezifikation
AAH32329 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.95kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KTLVQVAIYALGREPTFFFDPENFDPTRWLSKDKNITYFRNLGFGWGVRQCLGRRIAELEMTIFLINMLENFRVEIQHLSDVGTTFNLILMPEKPISFTFWPFNQEATQQ | |
RUO | |
CYP11A1 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
1583 | |
CYP11A1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CYP11A|P450SCC | |
CYP11A1 | |
Yes |