missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CXCL12, Recombinant Protein
E. coli Recombinant Protein
94.75 € - 361.00 €
Specifications
Accession Number | P48061 |
---|---|
For Use With (Application) | Control, ELISA, Immunohistochemistry, Western Blot |
Formulation | protein with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 |
Gene ID (Entrez) | 6387 |
Name | Human CXCL12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17134183
|
Invitrogen™
A42506 |
50 μg |
361.00 €
50µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17124183
|
Invitrogen™
A42505 |
5 μg |
94.75 €
5µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17114183
|
Invitrogen™
A42504 |
5 x 5 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
CXCL12 is a stromal cell-derived alpha chemokine member of the intercrine family. This gene product and its receptor CXCR4 can activate lymphocytes and have been implicated in the metastasis of some cancers such as breast cancer. Mutations in this gene are associated with resistance to human immunodeficiency virus type 1 infections. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
P48061 | |
protein with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | |
Human CXCL12 | |
-20°C | |
<1 EU/μg | |
CXCL12 | |
Unconjugated | |
Lyophilized |
Control, ELISA, Immunohistochemistry, Western Blot | |
6387 | |
E. coli | |
RUO | |
12-O-tetradecanoylphorbol 13-acetate repressed protein 1; AI174028; C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 12; chemokine (C-X-C motif) ligand 12 (stromal cell-derived factor 1); CXC; CXC motif chemokine; C-X-C motif chemokine 12; C-X-C motif chemokine ligand 12; CXCL; CXCL12; hIRH; hSDF-1; Intercrine reduced in hepatomas; IRH; Pbsf; Pre-B cell growth-stimulating factor; pre-B-cell growth-stimulating factor; SCYB12; Sdf1; SDF-1; SDF-1 gamma; SDF1A; SDF-1 A; SDF-1-alpha(3-67); SDF1B; SDF-1 b; SDF-1-beta(3-72); Stromal cell-derived factor 1; stromal cell-derived factor-1 alpha; stromal cell-derived factor-1 gamma; thymic lymphoma cell-stimulating factor; TLSF; Tlsfa; TLSF-a; Tlsfb; TLSF-b; Tpar1 | |
Human | |
Recombinant | |
>95% |