Learn More
Abnova™ Human CTHRC1 Partial ORF (NP_612464, 32 a.a. - 141 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00115908-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
CTHRC1 is specifically expressed in vascular calcifications of carotid artery lesions and may contribute to vascular remodeling of injured arteries (Pyagay et al., 2005 [PubMed 15618538]).[supplied by OMIM]
Sequence: EIPKGKQKAQLRQREVVDLYNGMCLQGPAGVPGRDGSPGANGIPGTPGIPGRDGFKGEKGECLRESFEESWTPNYKQCSWSSLNYGIDLGKIAECTFTKMRSNSALRVLFSpecifications
NP_612464 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EIPKGKQKAQLRQREVVDLYNGMCLQGPAGVPGRDGSPGANGIPGTPGIPGRDGFKGEKGECLRESFEESWTPNYKQCSWSSLNYGIDLGKIAECTFTKMRSNSALRVLF | |
RUO | |
CTHRC1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
115908 | |
CTHRC1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CTHRC1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |