Learn More
Abnova™ Human CPT2 Full-length ORF (AAH02445.1, 1 a.a. - 658 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00001376-P01.10ug
Weitere Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a nuclear protein which is transported to the mitochondrial inner membrane. Together with carnitine palmitoyltransferase I, the encoded protein oxidizes long-chain fatty acids in the mitochondria. Defects in this gene are associated with mitochondrial long-chain fatty-acid (LCFA) oxidation disorders. [provided by RefSeq]
Sequence: MVPRLLLRAWPRGPAVGPGAPSRPLSAGSGPGQYLQRSIVPTMHYQDSLPRLPIPKLEDTIRRYLSAQKPLLNDGQFRKTEQFCKSFENGIGKELHEQLVALDKQNKHTSYILGPWFDMYLSARDSVVLNFNPFMAFNPDPKSEYNDQLTRATNMTVSAIRFLKTLRAGLLEPEVFHLNPAKSDTITFKRLIRFVPSSLSWYGAYLVNAYPLDMSQYFRLFNSTRLPKPSRDELFTDDKARHLLVLRKGNFYIFDVLDQDGNIVSPSEIQAHLKYILSDSSPAPEFPLAYLTSENRDIWAELRQKLMSSGNEESLRKVDSAVFCLCLDDFPIKDLVHLSHNMLHGDGTNRWFDKSFNLIIAKDGSTAIHFEHSWGDGVAVLRFFNEVFKDSTQTPAVTPQSQPATTDSTVTVQKLNFELTDALKTGITAAKEKFDATMKTLTIDCVQFQRGGKEFLKKQKLSPDAVAQLAFQMAFLRQYGQTVATYESCSTAAFKHGRTETIRPASVYTKRCSEAFVREPSRHSAGELQQMMVECSKYHGQLTKEAAMGQGFDRHLFALRHLAAAKGIILPELYLDPAYGQINHNVLSTSTLSSPAVNLGGFAPVVSDGFGVGYAVHDNWIGCNVSSYPGRNAREFLQCVEKALEDVFDALEGKSIKSSpezifikation
AAH02445.1 | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
100.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CPT1|CPTASE | |
CPT2 | |
Yes |
wheat germ expression system | |
Liquid | |
1376 | |
CPT2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVPRLLLRAWPRGPAVGPGAPSRPLSAGSGPGQYLQRSIVPTMHYQDSLPRLPIPKLEDTIRRYLSAQKPLLNDGQFRKTEQFCKSFENGIGKELHEQLVALDKQNKHTSYILGPWFDMYLSARDSVVLNFNPFMAFNPDPKSEYNDQLTRATNMTVSAIRFLKTLRAGLLEPEVFHLNPAKSDTITFKRLIRFVPSSLSWYGAYLVNAYPLDMSQYFRLFNSTRLPKPSRDELFTDDKARHLLVLRKGNFYIFDVLDQDGNIVSPSEIQAHLKYILSDSSPAPEFPLAYLTSENRDIWAELRQKLMSSGNEESLRKVDSAVFCLCLDDFPIKDLVHLSHNMLHGDGTNRWFDKSFNLIIAKDGSTAIHFEHSWGDGVAVLRFFNEVFKDSTQTPAVTPQSQPATTDSTVTVQKLNFELTDALKTGITAAKEKFDATMKTLTIDCVQFQRGGKEFLKKQKLSPDAVAQLAFQMAFLRQYGQTVATYESCSTAAFKHGRTETIRPASVYTKRCSEAFVREPSRHSAGELQQMMVECSKYHGQLTKEAAMGQGFDRHLFALRHLAAAKGIILPELYLDPAYGQINHNVLSTSTLSSPAVNLGGFAPVVSDGFGVGYAVHDNWIGCNVSSYPGRNAREFLQCVEKALEDVFDALEGKSIKS | |
RUO | |
CPT2 | |
Wheat Germ (in vitro) | |
GST |