missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CPT2 Full-length ORF (AAH02445.1, 1 a.a. - 658 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
POA
Specifications
Name | CPT2 (Human) Recombinant Protein (P01) |
---|---|
Recombinant | Yes |
Purification Method | Glutathione Sepharose 4 Fast Flow |
Molecular Weight (g/mol) | 100.2kDa |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Description
The protein encoded by this gene is a nuclear protein which is transported to the mitochondrial inner membrane. Together with carnitine palmitoyltransferase I, the encoded protein oxidizes long-chain fatty acids in the mitochondria. Defects in this gene are associated with mitochondrial long-chain fatty-acid (LCFA) oxidation disorders. [provided by RefSeq]
Sequence: MVPRLLLRAWPRGPAVGPGAPSRPLSAGSGPGQYLQRSIVPTMHYQDSLPRLPIPKLEDTIRRYLSAQKPLLNDGQFRKTEQFCKSFENGIGKELHEQLVALDKQNKHTSYILGPWFDMYLSARDSVVLNFNPFMAFNPDPKSEYNDQLTRATNMTVSAIRFLKTLRAGLLEPEVFHLNPAKSDTITFKRLIRFVPSSLSWYGAYLVNAYPLDMSQYFRLFNSTRLPKPSRDELFTDDKARHLLVLRKGNFYIFDVLDQDGNIVSPSEIQAHLKYILSDSSPAPEFPLAYLTSENRDIWAELRQKLMSSGNEESLRKVDSAVFCLCLDDFPIKDLVHLSHNMLHGDGTNRWFDKSFNLIIAKDGSTAIHFEHSWGDGVAVLRFFNEVFKDSTQTPAVTPQSQPATTDSTVTVQKLNFELTDALKTGITAAKEKFDATMKTLTIDCVQFQRGGKEFLKKQKLSPDAVAQLAFQMAFLRQYGQTVATYESCSTAAFKHGRTETIRPASVYTKRCSEAFVREPSRHSAGELQQMMVECSKYHGQLTKEAAMGQGFDRHLFALRHLAAAKGIILPELYLDPAYGQINHNVLSTSTLSSPAVNLGGFAPVVSDGFGVGYAVHDNWIGCNVSSYPGRNAREFLQCVEKALEDVFDALEGKSIKSSpecifications
CPT2 (Human) Recombinant Protein (P01) | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Yes | |
100.2kDa |