Learn More
Abnova™ Human CPNE4 Partial ORF (NP_570720.1, 24 a.a. - 130 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00131034-Q03.10ug
Additional Details : Weight : 0.02000kg
Description
Calcium-dependent membrane-binding proteins may regulate molecular events at the interface of the cell membrane and cytoplasm. This gene is one of several genes that encode a calcium-dependent protein containing two N-terminal type II C2 domains and an integrin A domain-like sequence in the C-terminus. [provided by RefSeq]
Sequence: TKVELRVACKGISDRDALSKPDPCVILKMQSHGQWFEVDRTEVIRTCINPVYSKLFTVDFYFEEVQRLRFEVHDISSNHNGLKEADFLGGMECTLGQIVSQRKLSKSSpecifications
NP_570720.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.40kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TKVELRVACKGISDRDALSKPDPCVILKMQSHGQWFEVDRTEVIRTCINPVYSKLFTVDFYFEEVQRLRFEVHDISSNHNGLKEADFLGGMECTLGQIVSQRKLSKS | |
RUO | |
CPNE4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
131034 | |
CPNE4 (Human) Recombinant Protein (Q03) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
COPN4/CPN4/MGC15604 | |
CPNE4 | |
Recombinant | |
wheat germ expression system |