missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human COX4I2 Partial ORF (NP_115998, 21 a.a. - 104 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084701-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100857

  • 508.00 € / 25µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cytochrome c oxidase (COX), the terminal enzyme of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. It is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may be involved in the regulation and assembly of the complex. This nuclear gene encodes isoform 2 of subunit IV. Isoform 1 of subunit IV is encoded by a different gene, however, the two genes show a similar structural organization. Subunit IV is the largest nuclear encoded subunit which plays a pivotal role in COX regulation. [provided by RefSeq]

Sequence: MHSSEGTTRGGGKMSPYTNCYAQRYYPMPEEPFCTELNAEEQALKEKEKGSWTQLTHAEKVALYRLQFNETFAEMNRRSNEWKT
Specifications

Specifications

NP_115998
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.98kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MHSSEGTTRGGGKMSPYTNCYAQRYYPMPEEPFCTELNAEEQALKEKEKGSWTQLTHAEKVALYRLQFNETFAEMNRRSNEWKT
RUO
COX4I2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
84701
COX4I2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
COX4/COX4-2/COX4B/COX4L2/COXIV-2/dJ857M17.2
COX4I2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions