missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human COX18 Full-length ORF (NP_776188.1, 1 a.a. - 333 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_776188.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 285521 |
Molecular Weight (g/mol) | 63.5kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16100024
|
Abnova™
H00285521-P01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16110024
|
Abnova™
H00285521-P01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
COX18 encodes a cytochrome c oxidase (COX)-assembly protein. The S. cerevisiae Cox18 protein catalyzes the insertion of the Cox2 (MTCO2; MIM 516040) C-terminal tail into the mitochondrial inner membrane, an intermediate step in the assembly of complex IV of the mitochondrial respiratory chain (Sacconi et al., 2005 [PubMed 16212937]).[supplied by OMIM]
Sequence: MLCRLGGRWLRPLPALQLWARDLPLAPVPTSGAKRPTLPVWAVAPVSAVHANGWYEALAASSPVRVAEEVLLGVHAATGLPWWGSILLSTVALRGAVTLPLAAYQHYILAKVENLQPEIKTIARHLNQEVAVRANQLGWSKRDARLTYLKNMRRLISELYVRDNCHPFKATVLVWIQLPMWIFMSFALRNLSTGAAHSEGFSVQEQLATGGILWFPDLTAPDSTWILPISVGVINLLIVEICALQKIGMSRFQTYITYFVRAMSVLMIPIAATVPSSIVLYWLCSSFVGLSQNLLLRSPGFRQLCRIPSTKSDSETPYKDIFAAFNTKFISRKSpecifications
NP_776188.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
MLCRLGGRWLRPLPALQLWARDLPLAPVPTSGAKRPTLPVWAVAPVSAVHANGWYEALAASSPVRVAEEVLLGVHAATGLPWWGSILLSTVALRGAVTLPLAAYQHYILAKVENLQPEIKTIARHLNQEVAVRANQLGWSKRDARLTYLKNMRRLISELYVRDNCHPFKATVLVWIQLPMWIFMSFALRNLSTGAAHSEGFSVQEQLATGGILWFPDLTAPDSTWILPISVGVINLLIVEICALQKIGMSRFQTYITYFVRAMSVLMIPIAATVPSSIVLYWLCSSFVGLSQNLLLRSPGFRQLCRIPSTKSDSETPYKDIFAAFNTKFISRK | |
RUO | |
COX18 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
285521 | |
COX18 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
COX18HS/FLJ38991/MGC126733 | |
COX18 | |
Recombinant | |
wheat germ expression system |