missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human COMMD8 Full-length ORF (NP_060315.1, 1 a.a. - 183 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00054951-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MEPEEGTPLWRLQKLPAELGPQLLHKIIDGICGRAYPVYQDYHTVWESEEWMHVLEDIAKFFKAIVGKNLPDEEIFQQLNQLNSLHQETIMKCVKSRKDEIKQALSREIVAISSAQLQDFDWQVKLALSSDKIAALRMPLLSLHLDVKENGEVKPYSIEMSREELQNLIQSLEAANKVVLQLKSpecifications
NP_060315.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
47.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ20502 | |
COMMD8 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
54951 | |
COMMD8 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEPEEGTPLWRLQKLPAELGPQLLHKIIDGICGRAYPVYQDYHTVWESEEWMHVLEDIAKFFKAIVGKNLPDEEIFQQLNQLNSLHQETIMKCVKSRKDEIKQALSREIVAISSAQLQDFDWQVKLALSSDKIAALRMPLLSLHLDVKENGEVKPYSIEMSREELQNLIQSLEAANKVVLQLK | |
RUO | |
COMMD8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |