missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CNTNAP4 Partial ORF (NP_207837, 1145 a.a. - 1244 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_207837 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 85445 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16101037
|
Abnova™
H00085445-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16191027
|
Abnova™
H00085445-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene product belongs to the neurexin family, members of which function in the vertebrate nervous system as cell adhesion molecules and receptors. This protein, like other neurexin proteins, contains epidermal growth factor repeats and laminin G domains. In addition, it includes an F5/8 type C domain, discoidin/neuropilin- and fibrinogen-like domains, and thrombospondin N-terminal-like domains. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: VLGRILEHSDVDQETALAGAQGFTGCLSAVQLSHVAPLKAALHPSHPDPVTVTGHVTESSCMAQPGTDATSRERTHSFADHSGTIDDREPLANAIKSDSASpecifications
NP_207837 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CASPR4/KIAA1763 | |
CNTNAP4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
85445 | |
CNTNAP4 (Human) Recombinant Protein (Q01) | |
VLGRILEHSDVDQETALAGAQGFTGCLSAVQLSHVAPLKAALHPSHPDPVTVTGHVTESSCMAQPGTDATSRERTHSFADHSGTIDDREPLANAIKSDSA | |
RUO | |
CNTNAP4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |