missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CNGA4 Full-length ORF (NP_001032406.1, 1 a.a. - 575 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_001032406.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1262 |
Molecular Weight (g/mol) | 92.4kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16183341
|
Abnova™
H00001262-P01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16173341
|
Abnova™
H00001262-P01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CNGA4 is a modulatory subunit of vertebrate cyclic nucleotide-gated membrane channels that transduce odorant signals (Munger et al., 2001 [PubMed 11739959]).[supplied by OMIM]
Sequence: MSQDTKVKTTESSPPAPSKARKLLPVLDPSGDYYYWWLNTMVFPVMYNLIILVCRACFPDLQHGYLVAWLVLDYTSDLLYLLDMVVRFHTGFLEQGILVVDKGRISSRYVRTWSFFLDLASLMPTDVVYVRLGPHTPTLRLNRFLRAPRLFEAFDRTETRTAYPNAFRIAKLMLYIFVVIHWNSCLYFALSRYLGFGRDAWVYPDPAQPGFERLRRQYLYSFYFSTLILTTVGDTPPPAREEEYLFMVGDFLLAVMGFATIMGSMSSVIYNMNTADAAFYPDHALVKKYMKLQHVNRKLERRVIDWYQHLQINKKMTNEVAILQHLPERLRAEVAVSVHLSTLSRVQIFQNCEASLLEELVLKLQPQTYSPGEYVCRKGDIGQEMYIIREGQLAVVADDGITQYAVLGAGLYFGEISIINIKGNMSGNRRTANIKSLGYSDLFCLSKEDLREVLSEYPQAQTIMEEKGREILLKMNKLDVNAEAAEIALQEATESRLRGLDQQLDDLQTKFARLLAELESSALKIAYRIERLEWQTREWPMPEDLAEADDEGEPEEGTSKDEEGRASQEGPPGPESpecifications
NP_001032406.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
92.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSQDTKVKTTESSPPAPSKARKLLPVLDPSGDYYYWWLNTMVFPVMYNLIILVCRACFPDLQHGYLVAWLVLDYTSDLLYLLDMVVRFHTGFLEQGILVVDKGRISSRYVRTWSFFLDLASLMPTDVVYVRLGPHTPTLRLNRFLRAPRLFEAFDRTETRTAYPNAFRIAKLMLYIFVVIHWNSCLYFALSRYLGFGRDAWVYPDPAQPGFERLRRQYLYSFYFSTLILTTVGDTPPPAREEEYLFMVGDFLLAVMGFATIMGSMSSVIYNMNTADAAFYPDHALVKKYMKLQHVNRKLERRVIDWYQHLQINKKMTNEVAILQHLPERLRAEVAVSVHLSTLSRVQIFQNCEASLLEELVLKLQPQTYSPGEYVCRKGDIGQEMYIIREGQLAVVADDGITQYAVLGAGLYFGEISIINIKGNMSGNRRTANIKSLGYSDLFCLSKEDLREVLSEYPQAQTIMEEKGREILLKMNKLDVNAEAAEIALQEATESRLRGLDQQLDDLQTKFARLLAELESSALKIAYRIERLEWQTREWPMPEDLAEADDEGEPEEGTSKDEEGRASQEGPPGPE | |
RUO | |
CNGA4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
1262 | |
CNGA4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CNCA2/CNG5/CNGB2/MGC126168/MGC126169/OCNC2/OCNCBETA/OCNCb | |
CNGA4 | |
Recombinant | |
wheat germ expression system |