missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CIRH1A Partial ORF (NP_116219, 587 a.a. - 686 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084916-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100947

  • 518.16 € / 25µg

Estimated Shipment: 30-12-2021
Add to basket

Description

Description

This gene encodes a WD40-repeat-containing protein that is localized to the nucleolus. Mutation of this gene causes North American Indian childhood cirrhosis, a severe intrahepatic cholestasis that results in transient neonatal jaundice, and progresses to periportal fibrosis and cirrhosis in childhood and adolescence. [provided by RefSeq]

Sequence: SFHPKRPMHILLHDAYMFCIIDKSLPLPNDKTLLYNPFPPTNESDVIRRRTAHAFKISKIYKPLLFMDLLDERTLVAVERPLDDIIAQLPPPIKKKKFGT
Specifications

Specifications

NP_116219
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SFHPKRPMHILLHDAYMFCIIDKSLPLPNDKTLLYNPFPPTNESDVIRRRTAHAFKISKIYKPLLFMDLLDERTLVAVERPLDDIIAQLPPPIKKKKFGT
RUO
CIRH1A
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
84916
CIRH1A (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CIRHIN/FLJ14728/FLJ17146/KIAA1988/NAIC/TEX292
CIRH1A
Yes
Videos
SDS
Documents
Certificates
Promotions

Promotions